Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody (3C5-1A4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259692
Description
GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GRP78/HSPA5 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
Mouse | |
78 kDa | |
100 μg | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
3309 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
3C5-1A4 | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
HSPA5 | |
Full-length recombinant rat GRP78 | |
Protein G purified | |
RUO | |
Primary | |
Detects approx 33 kDa protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction