Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody (3C5-1A4), Novus Biologicals™

Mouse Monoclonal Antibody
$499.50
Specifications
Antigen | GRP78/HSPA5 |
---|---|
Clone | 3C5-1A4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GRP78/HSPA5 | |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
HSPA5 | |
IgG1 κ | |
Protein G purified | |
78 kDa |
3C5-1A4 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3309 | |
Full-length recombinant rat GRP78 | |
Primary | |
Detects approx 33 kDa protein. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title