Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP78/HSPA5 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB300520Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300520Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300520Z Supplier Novus Biologicals Supplier No. NB300520AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, Primate (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GRP78/HSPA5
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbols HSPA5
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of rat GRP78 (within residues 600-654). [Uniprot: P06761]
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 3309
Test Specificity Mouse and rat. Does not cross-react with Human or African Green Monkey (COS cells).
Target Species Human, Mouse, Rat, Primate (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.