Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP78/HSPA5 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NBP106274PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106274PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106274PE Supplier Novus Biologicals Supplier No. NBP106274PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRP78/HSPA5 Polyclonal antibody specifically detects GRP78/HSPA5 in Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep, Zebrafish samples. It is validated for Flow Cytometry.

Specifications

Antigen GRP78/HSPA5
Applications Flow Cytometry
Classification Polyclonal
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbols HSPA5
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot# P11021]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 3309
Target Species Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.