Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRWD1/WDR28 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | GRWD1/WDR28 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-0739
|
Bethyl Laboratories, Inc
A301577AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
GRWD1 is a component of the 50S and 80S preribosomal complexes and plays a role in ribosome biogenesis (Gratenstein et al., 2005 [PubMed 15885502]).Specifications
| GRWD1/WDR28 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| CDW4, GRWD, RRB1, WDR28, CUL4- and DDB1-associated WDR protein 4, WD repeat domain 28, glutamate rich WD repeat protein GRWD, glutamate-rich WD repeat-containing protein 1, regulator of ribosome biogenesis 1 homolog | |
| GRWD1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BQ67 | |
| 83743 | |
| Between 350 and 400 | |
| Primary | |
| 4° C | |
| GRWD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title