Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GS 6201

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective antagonist of adenosine A2B receptors (Ki values are 22, 1070, 1940 and 3280 nM for human A2B, A3, A1 and A2A receptors respectively). Attenuates allergen-, NECA- and AMP-induced airway reactivity in an allergic mouse model of asthma. Also shown to attenuate the inflammatory response during acute myocardial infarction in mice; reduces caspase-1 activity in the heart.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 446.43 |
| Purity | >99% |
| Molecular Weight (g/mol) | 446.434 |
| Chemical Name or Material | GS 6201 |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction