Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSDMDC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$416.50 - $706.00
Specifications
Antigen | GSDMDC1 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunocytochemistry/Immunofluorescence -Reactivity reported in scientific literature (PMID: 30250284)., Immunoprecipitation -Reported in scientific literature (PMID :33053349)., Immunohistochemistry-Paraffin -Reported in scientific literature (PMID:34494385)., Knockout Validated -Reactivity reported in scientific literature (PMID: 32796818)., Knockdown Validated -Reactivity reported in scientific literature (PMID: 31216481) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GSDMDC1 Polyclonal antibody specifically detects GSDMDC1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Knockdown.Specifications
GSDMDC1 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
DF5L, DFNA5L, FLJ12150, gasdermin D, gasdermin domain containing 1, Gasdermin domain-containing protein 1, gasdermin-D, GSDMDC1 | |
GSDMD | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04 - 0.4 ug/mL, Immunocytochemistry/Immunofluorescence -Reactivity reported in scientific literature (PMID: 30250284)., Immunoprecipitation -Reported in scientific literature (PMID :33053349)., Immunohistochemistry-Paraffin -Reported in scientific literature (PMID:34494385)., Knockout Validated -Reactivity reported in scientific literature (PMID: 32796818)., Knockdown Validated -Reactivity reported in scientific literature (PMID: 31216481) | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat, Pig | |
P57764 | |
79792 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GDNVYVVTEVLQTQKEVEVTRTHKREGSGRFSLPGATCLQGEGQGHLSQKKTVTIPSGSTLAFRVAQLVIDSDLDVLLFPDKKQRTFQPPATGHKRSTS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title