Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSDMDC1 Rabbit anti-Human, Clone: 8A8A5, Novus Biologicals™
SDP

Catalog No. NB167235 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167235 100 μg
NB167236 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167235 Supplier Novus Biologicals Supplier No. NBP316070100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSDMDC1 Monoclonal antibody specifically detects GSDMDC1 in Human samples. It is validated for Western Blot

Specifications

Antigen GSDMDC1
Applications Western Blot
Classification Monoclonal
Clone 8A8A5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DF5L, DFNA5L, FLJ12150, gasdermin D, gasdermin domain containing 1, Gasdermin domain-containing protein 1, gasdermin-D, GSDMD N, GSDMDC1, GSDMD-N
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 346-484 of human GSDMDC1 (NP_079012.3). DGPAGAVLECLVLSSGMLVPELAIPVVYLLGALTMLSETQHKLLAEALESQTLLGPLELVGSLLEQSAPWQERSTMSLPPGLLGNSWGEGAPAWVLLDECGLELGEDTPHVCWEPQAQGRMCALYASLALLSGLSQEPH
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 79792
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.