Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114023
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5114023 100 μL
1 options

Catalog No. PIPA5114023

Supplier: Invitrogen™ PA5114023

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NYWFVGTQKV PKPLCEKGLA AKCFDMPVSL DGDTNTSTQE VVQYNWETGD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 80%.

May cause the redistribution of PAPOLB from the cytosol to the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GSG1
Gene Accession No. Q2KHT4
Gene Alias germ cell associated 1; germ cell-specific gene 1 protein; GSG1; testicular secretory protein Li 20; UNQ709/PRO1360
Gene Symbols GSG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human GSG1 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83445
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.