Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 2334470
GREENER_CHOICE

Catalog No. 414310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-431-0 10 mg
41-435-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-431-0 Supplier Tocris Bioscience™ Supplier No. 4143/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent, selective PDPK1 (PDK1) inhibitor

Potent 3-phosphoinositide-dependent protein kinase (PDPK1) inhibitor (IC50 ∽ 10 nM). Exhibits no effect on other kinases including Aurora, ROCK, p38 MAPK and PI 3-K. Suppresses T-loop phosphorylation and subsequent activation of PDK1 substrates S6K1, SGK and RSK in vitro; exhibits limited inhibitory effect on Akt activation. Delays melanomagenesis and metastasis in BrafV600E::Pten(-/-) mice.

Chemical Identifiers

CAS 1227911-45-6
Molecular Formula C25H34N8O

Specifications

Quantity 10 mg
Formula Weight 462.59
Purity >98%
Molecular Weight (g/mol) 462.602
Chemical Name or Material GSK 2334470
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.