Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK 2606414

Click to view available options
Quantity:
10 mg
50 mg
Description
GSK 2606414 is a potent and selective protein kinase R-like ER kinase (PERK) inhibitor (IC50 = 0.4 nM). Exhibits >1000-fold selectivity for PERK over HR1 and PKR. Inhibits thapsigargin-induced PERK phosphorylation in lung carcinoma A549 cells. Attenuates subcutaneous pancreatic human tumor xenograft growth in mice. Orally bioavailable.
Specifications
Specifications
Quantity | 50 mg |
Product Type | GSK 2606414 |
Purity | >99% |
Molecular Weight (g/mol) | 451.44 |
Inhibitors | GSK 2606414 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction