Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 264220A
GREENER_CHOICE

Catalog No. 375210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-5210 10 mg
37-525-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-5210 Supplier Tocris Bioscience™ Supplier No. 3752/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endothelial lipase and lipoprotein lipase inhibitor

Endothelial lipase and lipoprotein lipase inhibitor (IC50 values are 0.13 and 0.10 μM respectively).

Chemical Identifiers

CAS 685506-42-7
Molecular Formula C17H21N3O4S

Specifications

Quantity 10 mg
Formula Weight 363.43
Purity >98%
Molecular Weight (g/mol) 363.432
Chemical Name or Material GSK 264220A
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.