Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 269962
GREENER_CHOICE

Catalog No. p-4816522
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective ROCK inhibitor

Potent Rho kinase (ROCK) inhibitor (IC50 values are 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively). Displays greater than 30-fold selectivity for ROCK against a panel of serine/threonine kinases. Induces vasorelaxation in preconstricted rat aorta (IC50 = 35 nM); lowers blood pressure in a rat model of hypertension.

Chemical Identifiers

CAS 850664-21-0
Molecular Formula C29H30N8O5

Specifications

Quantity 10 mg
Formula Weight 570.6
Purity >99%
Molecular Weight (g/mol) 570.61
Chemical Name or Material GSK 269962
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.