Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 alpha/beta Antibody (L5), Novus Biologicals™
SDP

Catalog No. NB019496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019496 0.025 mL
NBP229873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019496 Supplier Novus Biologicals Supplier No. NBP2298730.025ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen GSK-3 alpha/beta
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone L5
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation No buffer with No Preservative
Gene Accession No. P49840
Gene Alias DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha
Gene Symbols GSK3A
Host Species Rabbit
Immunogen N-terminal 6His-tagged recombinant full length human GSK3Beta.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2931
Test Specificity GSK3 alpha and beta 46kDa(beta)and51kDa(alpha) weight may be detected.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.