Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 beta, p Ser9 Antibody (3A6), Novus Biologicals™
SDP

Catalog No. NB298133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298133 100 μL
NB298132 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298133 Supplier Novus Biologicals Supplier No. NBP326422100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen GSK-3 beta
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 3A6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Host Species Rabbit
Immunogen A synthesized peptide derived from Human GSK-3 beta [p Ser 9] [UniProt P49841]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2932
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.