Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 beta, p Ser9 Antibody (609739), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB304391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304391 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304391 Supplier Novus Biologicals Supplier No. FAB25062G

Mouse Monoclonal Antibody

GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen GSK-3 beta
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 609739
Conjugate Alexa Fluor 488
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Host Species Mouse
Immunogen Phosphopeptide containing the human GSK-3 beta S9 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2932
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.