Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 beta, p Tyr279, p Tyr216 Antibody, Novus Biologicals™
SDP

Catalog No. NB10081946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10081946 0.05 mL
NB017843 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10081946 Supplier Novus Biologicals Supplier No. NB10081946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

GSK-3 beta (p Tyr216, p Tyr279) Polyclonal specifically detects GSK-3 beta (p Tyr216, p Tyr279) in Human, Mammalia, Mouse, Rat samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen GSK-3 beta (p Tyr216, p Tyr279)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. P49841
Gene Alias EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Gene Symbols GSK3B
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human GSK-3 beta around the phosphorylation site of tyrosine 279/216 (V-S-YP-I-C).
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2932
Test Specificity Detects endogenous levels of GSK-3 beta only when phosphorylated at tyrosine 216/279.
Target Species Human, Mouse, Mammalia, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.