Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK-3 beta, p Tyr279, p Tyr216 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$279.00 - $518.50
Specifications
Concentration | 1.0 mg/mL |
---|---|
Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017843
![]() |
Novus Biologicals
NB100819460.025ML |
0.025 mL |
Each for $279.00
|
|
|||||
NB10081946
![]() |
Novus Biologicals
NB10081946 |
0.05 mL |
Each for $518.50
|
|
|||||
Description
GSK-3 beta Polyclonal specifically detects GSK-3 beta in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Cancer, Hypoxia, Lipid and Metabolism, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
GSK3B | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
P49841 | |
2932 | |
IgG | |
Affinity Purified | |
46 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title