Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3alpha/beta Rabbit anti-Human, Clone: SY26-05, Novus Biologicals™
SDP

Catalog No. NBP267441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267441 Supplier Novus Biologicals Supplier No. NBP267441
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen GSK-3alpha/beta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SY26-05
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:800
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha
Gene Symbols GSK3A
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Tyr216 and 279 of human GSK-3 alpha/beta (SwissProt: P49840 Human; SwissProt: P49841 Human; SwissProt: Q2NL51 Mouse; SwissProt: Q9WV60 Mouse; SwissProt: P18265 Rat; SwissProt: P18266 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2931
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.