Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 583
GREENER_CHOICE

Catalog No. p-7146002
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent RIP2 kinase inhibitor; orally bioavailable and cell permeable

GSK 583 is a potent RIPK2 inhibitor (IC50 values are 2 and 5 nM at rat and human kinases, respectively). Displays selectivity for RIP2K over p38α, VEGFR2 and a panel of 300 kinases. Exhibits some inhibition at BRK and Aurora A. Also displays similar binding affinity, but no detectable functional activity at RIP3. Active in vivo. Moderately orally bioavailable and cell permeable.

Specifications

Chemical Name or Material 6-[(1,1-Dimethylethyl)sulfonyl]-N-(5-fluoro-1H-indazol-3-yl)-4-quinolinamine
CAS 1346547-00-9
Quantity 5 mg
Molecular Formula C20H19FN4O2S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.