Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 650394
GREENER_CHOICE

Catalog No. 357250R
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Serum- and glucocorticoid-regulated kinase (SGK) inhibitor

GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).

Specifications

Quantity 50 mg
Product Type GSK 650394
Purity >97%
Molecular Weight (g/mol) 382.45
Inhibitors GSK 650394

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.