Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 6853
GREENER_CHOICE

Catalog No. p-7161629
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective BRPF1 inhibitor

GSK 6853 is a potent and selective BRPF1 inhibitor. Exhibits >1600-fold selectivity for BRPF1 over other bromodomains (pIC50 values by TR-FRET are 8.1, 5.1/4.8 and 4.7/<4.3 for BRPF1, BRPF2/3 and BRD4 BD1/2, respectively).

Negative Control also available.

Specifications

Chemical Name or Material N-[2,3-dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide
CAS 1910124-24-1
Quantity 25 mg
Molecular Formula C22H27N5O3
Formula Weight 409.48
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.