Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK'872

Click to view available options
Quantity:
10 mg
50 mg
Description
GSK'872 is a potent and selective receptor activating protein kinase 3 (RIP3) inhibitor (IC50 = 1.3 nM). Exhibits >1000-fold selectivity for RIP3 over a panel of 300 other kinases including RIP1. Inhibits TNF-induced cell death in 3T3-SA cells and TLR3-induced necrosis in fibroblasts in vitro. Blocks necroptosis of primary human neutrophils from whole blood. Also induces apoptosis at higher concentrations (3 - 10 μM).
Specifications
Specifications
| Chemical Name or Material | N-5-Benzothiazolyl-6-[(1-methylethyl)sulfonyl]-4-quinolinamine |
| CAS | 1346546-69-7 |
| Quantity | 10 mg |
| Molecular Formula | C19H17N3O2S2 |
| Formula Weight | 383.49 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction