Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK3A Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | GSK3A |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8730
![]()
|
Proteintech Group Inc
134191AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Glycogen Synthase Kinase-3 differs from most serine/threonine kinases in that it is active in the absence of the action of signaling pathways. Two isoforms exist, GSK-3alpha and GSK-3beta. The function of GSK-3 is to phosphorylate glycogen synthase and thereby inactivate it. Insulin action stimulates the PI 3-kinase pathway, resulting in Akt activation, which phosphorylates and inactivates GSK-3.Specifications
| GSK3A | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Zebrafish | |
| P18265, P49840, Q2NL51 | |
| 2931, 30664, 50686, 606496 | |
| GSK3A Fusion Protein Ag4168 | |
| Primary | |
| -20°C | |
| GSK3A |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GSK3 alpha, GSK3A | |
| Gsk3a, gsk3ab | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title