Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSPT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15764620UL
Description
GSPT2 Polyclonal specifically detects GSPT2 in Human samples. It is validated for Western Blot.Specifications
| GSPT2 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Q8IYD1 | |
| GSPT2 | |
| Synthetic peptides corresponding to GSPT2 (G1 to S phase transition 2) The peptide sequence was selected from the middle region of GSPT2)(50ug). Peptide sequence GANIKEQSDFCPWYTGLPFIPYLDNLPNFNRSIDGPIRLPIVDKYKDMGT. | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 23708 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| eRF3b, ERF3BGST2, eukaryotic peptide chain release factor GTP-binding subunit ERF3B, Eukaryotic peptide chain release factor subunit 3b, FLJ10441, G1 to S phase transition 2, G1 to S phase transition protein 2 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction