Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Antibody (1E10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP250415IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250415IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250415IR Supplier Novus Biologicals Supplier No. NBP250415IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot.

Specifications

Antigen GST Epitope Tag
Applications Western Blot
Classification Monoclonal
Clone 10000000000
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Gene Symbols GSTM1
Host Species Mouse
Immunogen GST protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Epitope Tags, GST Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.