Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Antibody (3G10), Novus Biologicals™
SDP

Catalog No. NB600446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600446 Supplier Novus Biologicals Supplier No. NB600446
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen GST Epitope Tag
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 3G10
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-2 ug/ml, Immunoprecipitation 1:10 - 1:500
Gene Accession No. P09488
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Gene Symbols GSTM1
Host Species Mouse
Immunogen GST protein of the parasitic helminth Schistosoma japonicum expressed in E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Epitope Tags, GST Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Test Specificity Recognizes Glutathione S-Transferase (GST), a 26-kDa protein encoded by the parasitic helminth Schistosoma japonicum.
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.