Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Antibody (6I7V6), Novus Biologicals™
SDP

Catalog No. NB168862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB168862 Supplier Novus Biologicals Supplier No. NBP316888
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen GST Epitope Tag
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 6I7V6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-239 of schja GST protein (P08515).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Asthma, Cancer, Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.