Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB60325A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60325A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60325A750 Supplier Novus Biologicals Supplier No. NB600325AF750
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GST Epitope Tag Polyclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen GST Epitope Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Gene Symbols GSTM1
Host Species Goat
Immunogen Goats were immunized with Glutathione-S-Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2944
Content And Storage Store at 4°C in the dark.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.