Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB355536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355536 Supplier Novus Biologicals Supplier No. NBP237820PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GST Epitope Tag Polyclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot

Specifications

Antigen GST Epitope Tag
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Host Species Rabbit
Immunogen Purified recombinant GST.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.