Learn More
GST Monoclonal Antibody (2F10B9), THE™, GenScript™
Mouse Monoclonal Antibody
$183.60 - $790.50
Specifications
Antigen | GST |
---|---|
Clone | 2F10B9 |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-272-995
![]() |
Genscript Corporation
A00865100 |
100 μg |
Each for $183.60
|
|
|||||
50-272-997
![]() |
Genscript Corporation
A00865200 |
200 μg |
Each for $326.40
|
|
|||||
50-272-998
![]() |
Genscript Corporation
A00865500 |
500 μg |
Each for $790.50
|
|
|||||
Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Schistosoma japonicum glutathione S-transferase (GST) is a 26,000 Da enzyme that conjugates reduced glutathione to hydrophobic electrophiles. GST is an ideal fusion partner for production of foreign proteins in commercially available expression systems because it can be expressed in high levels in E. coli. The fusion proteins can be purified to homogenity in a single step as the GST portion of the protein binds tightly to immobilized glutathione. Elution can be accomplished using free glutathione. The GST portion of the fusion protein can be separated from the protein of interest by using site-specific proteases. Antibodies directed against GST can be helpful in detecting the fusion protein during purification and to detect cleavage of GST from the protein of interest.
GST Monoclonal antibody specifically detects GST in Tag samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
GST | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide; pH 7.4 | |
Recombinant GST protein (26 kD) | |
Primary | |
-20°C or -80°C if preferred |
2F10B9 | |
Monoclonal | |
Lyophilized | |
RUO | |
activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19 | |
IgG1 κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.