Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTA1 Monoclonal Antibody (2-2#), Invitrogen™

Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
| Antigen | GSTA1 |
|---|---|
| Clone | 4# |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543789
|
Invitrogen™
MA543789 |
20 μL |
Each for $213.50
|
|
|||||
PIMA543788
![]() |
Invitrogen™
MA543788 |
100 μL |
Each for $522.50
|
|
|||||
Description
This gene encodes a member of a family of enzymes that function to add glutathione to target electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress. This action is an important step in detoxification of these compounds. This subfamily of enzymes has a particular role in protecting cells from reactive oxygen species and the products of peroxidation. Polymorphisms in this gene influence the ability of individuals to metabolize different drugs. This gene is located in a cluster of similar genes and pseudogenes on chromosome 6. Alternative splicing results in multiple transcript variants.Specifications
| GSTA1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; class alpha; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase; glutathione S-transferase 2; glutathione S-transferase A1; Glutathione S-transferase A1, N-terminally processed; glutathione S-transferase A3 subunit; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha M14; Glutathione S-transferase alpha M14, N-terminally processed; Glutathione S-transferase alpha-3; glutathione S-transferase Ha subunit 1; glutathione S-transferase Ya; Glutathione S-transferase Ya1; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; glutathione S-transferase, alpha 1 (Ya); Glutathione-S-transferase alpha type (Ya); GST; GST 2-2; GST A3-3; GST AA; GST class-alpha; GST class-alpha member 1; GST HA subunit 1; GST Yc1; GST2; Gst2-1; Gsta; GSTA1; GSTA1-1; Gsta3; Gsta5; GST-epsilon; Gstya; Gstyc1; GTH1; OTTMUSG00000031890; RP11-392A23.1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80; Yc1 | |
| GSTA1 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| 4# | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08263 | |
| 2938 | |
| Ala2-Phe222 (Accession P08263), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GSTA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title