Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTA4 Antibody (OTI3F5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270856A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270856A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270856A48 Supplier Novus Biologicals Supplier No. NBP270856AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTA4 Monoclonal specifically detects GSTA4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen GSTA4
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3F5
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4
Gene Symbols GSTA4
Host Species Mouse
Immunogen Full length recombinant protein of human GSTA4 (NP_001503) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2941.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.