Learn More
Description
Specifications
Specifications
| Antigen | GSTA4 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI3F5 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4 |
| Gene Symbols | GSTA4 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
