Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTA4 Antibody (SR1360), Novus Biologicals™
SDP

Catalog No. NB260307 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260307 50 μg
NB258160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260307 Supplier Novus Biologicals Supplier No. NBP32264650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSTA4 Monoclonal antibody specifically detects GSTA4 in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen GSTA4
Applications Western Blot
Classification Monoclonal
Clone SR1360
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSTA4 (Uniprot #: O15217)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2941
Target Species Human, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.