Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSTA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | GSTA5 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119148
|
Invitrogen™
PA5119148 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121940
|
Invitrogen™
PA5121940 |
20 μL |
Each for $237.00
|
|
|||||
Description
GSTA5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The glutathione S-transferases catalyze the conjugation of reduced glutathiones and a variety of electrophiles, including many known carcinogens and mutagens. The cytosolic GSTs belong to a large superfamily, with members located on different chromosomes.Specifications
GSTA5 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Rat | |
GSTA5 | |
glutathione S-transferase A5; Glutathione S-transferase A5-5; glutathione S-transferase alpha 5; glutathione transferase A5; GST class-alpha member 5; GSTA5 | |
GSTA5 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P46418, Q7RTV2 | |
221357, 494499 | |
Recombinant protein Glutathione S Transferase Alpha 5. The antigen corresponds to amino acid range 1-221 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title