Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTM2 Antibody (CPTC-GSTMu2-2), PerCP, Novus Biologicals™
SDP

Catalog No. NB382326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382326 Supplier Novus Biologicals Supplier No. NBP308255PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTM2 Monoclonal antibody specifically detects GSTM2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GSTM2
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-GSTMu2-2
Conjugate PerCP
Formulation PBS
Gene Alias glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2
Host Species Mouse
Immunogen Recombinant human full-length GSTM2 protein (Uniprot: P28161)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2946
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.