Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTO1 Antibody (14A9.F6), Biotin, Novus Biologicals™
SDP

Catalog No. NB120874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120874 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120874 Supplier Novus Biologicals Supplier No. NBP244176B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTO1 Monoclonal antibody specifically detects GSTO1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen GSTO1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 14A9.F6
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias DKFZp686H13163, EC 2.5.1.18, glutathione S-transferase omega 1, glutathione S-transferase omega 1-1, glutathione S-transferase omega-1, glutathione-S-transferase like, GSTO-1, GSTTLp28, P28
Host Species Mouse
Immunogen This Protein A purified GSTO1 Antibody (14A9.F6) was prepared by repeated immunizations in mice with GSTGSTO1 Protein (Uniprot: P78417)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9446
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.