Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTO2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15458020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15458020 20 μL
NBP154580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15458020 Supplier Novus Biologicals Supplier No. NBP15458020UL

Rabbit Polyclonal Antibody

GSTO2 Polyclonal specifically detects GSTO2 in Human samples. It is validated for Western Blot.

Specifications

Antigen GSTO2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9H4Y5
Gene Alias bA127L20.1, bA127L20.1 (novel glutathione-S-transferase), EC 2.5.1.18, glutathione S-transferase omega 2, glutathione S-transferase omega-2, glutathione-S-transferase-like protein, GSTO-2
Gene Symbols GSTO2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GSTO2(glutathione S-transferase omega 2) The peptide sequence was selected from the N terminal of GSTO2. Peptide sequence VLETSQCQLIYESVIACEYLDDAYPGRKLFPYDPYERARQKMLLELFCKV.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119391
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.