Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTT2 Antibody (AT10H4), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP250577C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250577C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250577C Supplier Novus Biologicals Supplier No. NBP250577C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTT2 Monoclonal specifically detects GSTT2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GSTT2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AT10H4
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.5.1.18, glutathione S-transferase theta 2, glutathione S-transferase theta-2, Glutathione S-transferase theta-2B, GST class-theta-2, GSTT2B, MGC182032
Gene Symbols GSTT2
Host Species Mouse
Immunogen Recombinant human GSTT2 (1-244aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2953
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.