Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTZ1 Antibody (044), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB333995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333995 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333995 Supplier Novus Biologicals Supplier No. NBP290273MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human samples. It is validated for ELISA

Specifications

Antigen GSTZ1
Applications ELISA
Classification Monoclonal
Clone 044
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human GSTZ1 (Accession#: NP_665877.1, Met1-Ala216)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2954
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.