Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTZ1 Antibody (24E9.G9.G8), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB120889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120889 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120889 Supplier Novus Biologicals Supplier No. NBP244177AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen GSTZ1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 24E9.G9.G8
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase
Host Species Mouse
Immunogen This Protein A purified GSTZ1 Antibody (24E9.G9.G8) was prepared by repeated immunizations in mice with GSTGSTZ1 Protein (Uniprot: O43708)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2954
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.