Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2A1 Antibody (PCRP-GTF2A1-1F2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB187269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187269 Supplier Novus Biologicals Supplier No. NBP314054JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GTF2A1 Monoclonal antibody specifically detects GTF2A1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen GTF2A1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-GTF2A1-1F2
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias General transcription factor IIA subunit 1, general transcription factor IIA, 1, 19/37kDa, glucose regulated protein, 58kD pseudogene, MGC129969, MGC129970, TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, transcription initiation factor IIA subunit 1, Transcription initiation factor TFIIA 42 kDa subunit
Host Species Mouse
Immunogen Recombinant full-length human GTF2A1 protein (Uniprot: P52655)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2957
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.