Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2H2C Antibody (PCRP-GTF2H2C-2C9), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB389647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389647 Supplier Novus Biologicals Supplier No. NBP308896JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GTF2H2C Monoclonal antibody specifically detects GTF2H2C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen GTF2H2C
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-GTF2H2C-2C9
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias General transcription factor IIH polypeptide 2-like protein, general transcription factor IIH subunit 2-like protein, general transcription factor IIH, polypeptide 2C, GTF2H2C_2
Host Species Mouse
Immunogen Recombinant full-length human GTF2H2C protein (Uniprot: Q6P1K8)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 728340
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.