Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD2 Antibody (PCRP-GTF2IRD2-1B12), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB386397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386397 Supplier Novus Biologicals Supplier No. NBP308512FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GTF2IRD2 Monoclonal antibody specifically detects GTF2IRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array

Specifications

Antigen GTF2IRD2
Applications Western Blot, Flow Cytometry, Protein Array
Classification Monoclonal
Clone PCRP-GTF2IRD2-1B12
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha
Host Species Mouse
Immunogen Recombinant full-length human GTF2IRD2 protein (Uniprot: Q86UP8)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84163
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.