Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPBP9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216768 Supplier Novus Biologicals Supplier No. NBP216768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTPBP9 Polyclonal specifically detects GTPBP9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Tumorsphere.

Specifications

Antigen GTPBP9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.46mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9NTK5
Gene Alias DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004
Gene Symbols OLA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GTPBP9. The exact sequence is proprietary.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 29789
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Chicken (90%), Rat (99%), Xenopus laevis (85%), Zebrafish (89%), Bovine (98%), Xenopus tropicalis (87%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.