Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPBP9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP157545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP157545 Supplier Novus Biologicals Supplier No. NBP157545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTPBP9 Polyclonal specifically detects GTPBP9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GTPBP9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NTK5
Gene Alias DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004
Gene Symbols OLA1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GTPBP9 (GTP-binding protein 9 (putative)) The peptide sequence was selected from the N terminal of GTPBP9. Peptide sequence QGLGNAFLSHISACDGIFHLTRAFEDDDITHVEGSVDPIRDIEIIHEELQ.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 29789
Test Specificity Expected identity based on immunogen sequence: Rat: 100%; Human: 100%; Chicken: 100%; Sumatran orangutan: 100%; Bovine: 100%; Mouse: 100%; Endeis laevis: 92%; Nicoletia meinerti: 92%; Skogsbergia lerneri: 92%; Streptocephalus seali: 92%; Brucella sp. 83/13: 92%; Brucella pinnipedialis M292/94/1: 92%; Brucella melitensis bv. 1 str. Rev.1: 92%; Brucella melitensis bv. 3 str. Ether: 92%; Woodlice: 92%; Brine shrimp: 92%; Ammothea hilgendorfi: 92%; Brucella abortus NCTC 8038: 92%; Brucella melitensis bv. 1 str. 16M: 92%; Brucella suis bv. 4 str. 40: 92%; Brucella melitensis bv. 2 str. 63/9: 92%; Brucella suis bv. 5 str. 513: 92%; Brucella suis bv. 3 str. 686: 92%; Brucella sp. F5/99: 92%; Achelia echinata: 92%; Brucella ceti M490/95/1: 92%; Gluconacetobacter hansenii ATCC 23769: 92%; Brucella sp. BO2: 92%; Sphingomonas sp. SKA58: 92%; Brucella abortus: 92%; Gluconobacter suboxydans: 92%; Zymomonas mobilis: 92%; Brucella suis: 92%; Brucella melitensis: 92%; Sinorhizobium meliloti: 92%; Harpegnathos saltator: 92%; Ahrensia sp. R2A130: 92%; Brucella sp. NVSL 07-0026: 92%; Red flour beetle: 92%; Brucella abortus bv. 5 str. B3196: 92%;.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.