Learn More
Guaiazulene, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
It is employed as antimicrobial agent and perfuming agent in cosmetics industry.
Solubility
Soluble in alcohol, water, 0.1115 mg/L @ 25°C (est).
Notes
Store in dark. Protect from heat, oxidizing agents and heat. Keep it refrigerated.
Specifications
Specifications
| Melting Point | 31°C to 33°C |
| Density | 0.976 |
| Boiling Point | 152°C to 154°C (7 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| Beilstein | 1365001 |
| Sensitivity | Light sensitive |
| Merck Index | 14,4554 |
| Solubility Information | Soluble in alcohol,water,0.1115mg/L @ 25°C (est). |
| Formula Weight | 198.31 |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 207-701-2 |
| RTECSNumber | CO4790000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.