Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiazulene, 99%

Catalog No. p-4228708
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

CAS: 489-84-9 | C15H18 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489-84-9
Molecular Formula C15H18
Molecular Weight (g/mol) 198.31
InChI Key FWKQNCXZGNBPFD-UHFFFAOYSA-N
Synonym guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
PubChem CID 3515
ChEBI CHEBI:5550
IUPAC Name 1,4-dimethyl-7-propan-2-ylazulene
SMILES CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

Specifications

Melting Point 29.0°C to 31.0°C
Color Blue
Density 0.9700g/mL
Boiling Point 153.0°C (7.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05,II,473
Merck Index 15,459
Specific Gravity 0.97
Solubility Information Solubility in water: 25%. Other solubilities: soluble in ethanol, ethyl ether and ethyl acetate
Formula Weight 198.31
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Guaiazulene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 207-701-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.