Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine, 98%

Catalog No. AAA1202422
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 73-40-5 | C5H5N5O | 151.13 g/mol

Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

Solubility
Insoluble in water. Soluble in ammonia-water, KOH solutions, dilute acids, and 1 N NaOH (0.1M).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 9680
Merck Index 14,4564
Solubility Information Insoluble in water. Soluble in ammonia-water,KOH solutions,dilute acids,and 1 N NaOH (0.1M).
IUPAC Name 2-amino-3,7-dihydropurin-6-one
Formula Weight 151.13
Percent Purity 98%
Chemical Name or Material Guanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-799-8
RTECSNumber MF8260000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.